Polyethylene glycol monomethyl ether, M.W. ∼ 750
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9004-74-4
:
Formula:CH₃(OCH₂CH₂)nOH
MDL Number:MFCD00084416
Density:1.09 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Solid or slightly hazy liquid
- Infrared spectrum:Conforms
- Average molecular weight:715 to 785
- pH:4.5 to 7.5 (5 % aq. soln., 25°C)