
Poly(2-acrylamido-2-methyl-1-propanesulphonic acid), M.W. ∼ 800,000 10% (w/w) in aqueous solution
:27119-07-9
:
:MFCD00084369
:Ambient
:3265
:8,III
:GHS05
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Assay:8.0 to 12.0 % Solids
- Solubility:(0.1% in ethanol) Clear colorless
- HPLC:≥96.0 %