



Lithium tri-sec-butylborohydride 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38721-52-7
:
Formula:C₁₂H₂₇BLi
MDL Number:MFCD00011708
:189.098 g/mol
Density:0.89 g/cm³
Flash Pt:–17 °C
UN:3129
ADR:4.3,I
:GHS05
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear to hazy colorless to yellow Solution
- Strength:0.80 to 1.10 M
- NMR (1H-NMR):Consistent with structure
- Elemental Analysis:86.23 to 88.85 % Carbon