Silicone oil high temperature
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63148-58-3
:63148-62-9
MDL Number:MFCD00132673
Density:1.07 g/cm³
Flash Pt:250 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to straw yellow Oily liquid
- Infrared spectrum:Conforms
- Refractive index:1.4940 to 1.5040 (20°C, 589 nm)
- Viscosity:100 to 150 cSt (at 25°C)