1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
ACRO121830050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
- Appearance:White to almost white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C)
- Titration with NaOH:≥97.5 % (on dried substance)
: Allopurinol
:
:136.11 g/mol
:384 °C
:Ambient
:2811
:6.1,III
:GHS06
:315-30-0
:C₅H₄N₄O
:MFCD00599413