3-Ethoxy-2-cyclohexen-1-one 99%
:5323-87-5
:
:C₈H₁₂O₂
:MFCD00001580
:140.18 g/mol
:76…78 °C (1 mmHg)
:1.04 g/cm³
:107 °C
:Refrigerator
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5020 to 1.5060 (20°C, 589 nm)
- GC:≥98.5 %
- Insoluble matter:≤0.005 %