Order Entry
China
ContactUsLinkComponent
3,3'-Diaminobenzidine tetrahydrochloride hydrate, (max. 10% H₂O) 97%
Health
3,3'-Diaminobenzidine tetrahydrochloride hydrate, (max. 10% H₂O) 97%
Certificates
undefined
Health
3,3'-Diaminobenzidine tetrahydrochloride hydrate, (max. 10% H₂O) 97%
Specification Test Results
  • Appearance:
    Grey to brown-purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥96.0 % (on dry substance)
  • Loss on drying:
    ≤10 % (120°C)