Order Entry
China
ContactUsLinkComponent
2-Isopropyl-N,2,3-trimethylbutyramide
Irritant
2-Isopropyl-N,2,3-trimethylbutyramide
Certificates
undefined
Irritant
2-Isopropyl-N,2,3-trimethylbutyramide

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    60 to 65 °C
  • Water:
    ≤0.5 %
  • GC:
    ≥99.4 %

Specification Test Results

Related Information