N-Benzoyl-N-phenylhydroxylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:304-88-1
:
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD00002111
:213.24 g/mol
Melting Pt:119…123 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light beige Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:119 to 123 °C
- HPLC:≥97.5 %