Order Entry
China
ContactUsLinkComponent
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%
Toxic
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%
Catalog # ACRO414020100
CAS Number:  
undefined
Toxic
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%
Catalog # ACRO414020100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤8 %
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :233.714 g/mol
Melting Pt:270…276 °C (dec.)
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:38894-11-0
Formula:C₈H₉N₃S·H₂O·HCl
MDL Number:MFCD00149370