Order Entry
China
ContactUsLinkComponent
 
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulphonate monohydrate 80% tech.
目录编号 ACRO400840050
CAS Number:  
undefined
Sodium hydrogen 4-amino-5-hydroxynaphthalene-2,7-disulphonate monohydrate 80% tech.
目录编号 ACRO400840050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Beige to grey Powder and/or lumps
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    4 % to 6 % (1 g, 150°C, 4 hrs)
  • HPLC:
    ≥75 %

Specification Test Results

About this item

 :
 :359.30 g/mol
Storage Temperature:Ambient
CAS Number:312693-54-2
 :343321-59-5
Formula:C₁₀H₈NNaO₇S₂·1H₂O
MDL Number:MFCD00150460