Diethyl (3,3-diethoxypropyl)phosphonate 95% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15110-17-5
:
Formula:C₁₁H₂₅O₅P
MDL Number:MFCD00800259
:max. 1% w/w Stabilised with sodium carbonate (Na₂CO₃)
Molecular Weight:268.29 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear light yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4320 (20°C, 589 nm)
- GC:≥94 %
- Stabilizer:≤1 % Na2CO3