Metacycline hydrochloride, 98%, sec. Standard
Catalog # ACRO303861000
CAS 编号:
规格
- Pack type:Glass bottle
- Pk:100 mg
- CAS No.:3963-95-9
- Formula:C₂₂H₂₂N₂O₈.HCl
- Description:Metacycline hydrochloride
规格
关于此项目
MDL No.: MFCD04972012
Appearance: Yellow Crystalline powder
Storage Temperature: Ambient