Order Entry
China
ContactUsLinkComponent
1,3,6-Naphthalenetrisulphonic acid, sodium salt hydrate mixture of isomers
Irritant
1,3,6-Naphthalenetrisulphonic acid, sodium salt hydrate mixture of isomers
Catalog # ACRO269010010
CAS Number:  
undefined
Irritant
1,3,6-Naphthalenetrisulphonic acid, sodium salt hydrate mixture of isomers
Catalog # ACRO269010010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance:
    White to yellow to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤12 %
  • HPLC:
    ≥90 % (mix of isomers)
  • Sodium sulfate (Na2SO4):
    ≤5

Specification Test Results

About this item

 :
 :390.34 g/mol (anhy)
 :GHS07
Storage Temperature:Ambient
CAS Number:19437-42-4
 :123409-01-8
Formula:C₁₀H₇NaO₉S₃·xH₂O