
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
Catalog # ACRO264110010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4700 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:+130° to +134° (20°C, 589 nm) (c=6, CHCl3)
Specification Test Results
About this item
:
:252.62 g/mol
Boiling Pt:213…214 °C (760 mmHg)
Density:1.34 g/cm³
Flash Pt:89 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
CAS Number:20445-33-4
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD00067105