beta-Apo-oxytetracycline, can be used as secondary standard
Catalog # ACRO257731000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- CAS No.:18751-99-0
- Formula:C₂₂H₂₂N₂O₈
- Description:beta-Apo-oxytetracycline
Specifications
About this item
CAS No.: 18751-99-0
MDL No.: MFCD00062827
GHS Symbols: GHS08
Melting Point: 200 to 204 °C (D)
Appearance: Yellow-brown to brown Powder
Storage Temperature: Ambient
Signal word: Warning