Order Entry
China
ContactUsLinkComponent
 
alpha-Apo-oxytetracycline, can be used as secondary standard
  ACRO257721000
 :  
alpha-Apo-oxytetracycline, can be used as secondary standard
  ACRO257721000
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • CAS No.:
    18695-01-7
  • Formula:
    C₂₂H₂₂N₂O₈
  • Description:
    alpha-Apo-oxytetracycline

 

 

CAS No.: 18695-01-7

MDL No.: MFCD00062827
GHS Symbols: GHS07, GHS08,
Melting Point: 200 °C (D)
Appearance: Khaki Crystalline powder
Storage Temperature: Ambient
Signal word: Warning