4-Epioxytetracycline, 97%, can be used as secondary standard
目录编号 ACRO257711000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- CAS No.:14206-58-7
- Formula:C₂₂H₂₄N₂O₉
- Description:4-Epioxytetracycline
Specifications
About this item
CAS No.: 14206-58-7
MDL No.: MFCD00021181
GHS Symbols: GHS07, GHS08,
Melting Point: 168 °C (D)
Appearance: Green-beige to brown Powder
Storage Temperature: Ambient
Signal word: Warning