Oxytetracycline, can be used as secondary standard
Catalog # ACRO257705000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- CAS No.:79-57-2
- Formula:C₂₂H₂₄N₂O₉
- Description:Oxytetracycline
Specifications
About this item
CAS No.: 79-57-2
MDL No.: MFCD00003700
GHS Symbols: GHS07, GHS08,
Melting Point: 183 °C (D)
Appearance: Light yellow to beige Crystalline powder
Storage Temperature: Ambient
Signal word: Warning