Order Entry
China
ContactUsLinkComponent
Dimethyl-cis-1,2-cyclopropanedicarboxylate 97+%
Irritant
Dimethyl-cis-1,2-cyclopropanedicarboxylate 97+%
Certificates
undefined
Irritant
Dimethyl-cis-1,2-cyclopropanedicarboxylate 97+%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4450 to 1.4470 (20°C, 589 nm)
  • GC:
    ≥97.0 %

Specification Test Results

Related Information