Order Entry
China
ContactUsLinkComponent
Anhydrotetracycline hydrochloride, can be used as a secondary standard
Anhydrotetracycline hydrochloride, can be used as a secondary standard
Certificates
undefined
Anhydrotetracycline hydrochloride, can be used as a secondary standard
Certificates

Specifications

  • CAS No.:
    13803-65-1
  • Formula:
    C₂₂H₂₂N₂O₇.HCl
  • Description:
    Anhydrotetracycline hydrochloride
  • Appearance:
    Yellow Powder

Specifications

Related Information