Order Entry
China
ContactUsLinkComponent
4-Epitetracycline hydrochloride, can be used as a secondary standard
4-Epitetracycline hydrochloride, can be used as a secondary standard
Certificates
4-Epitetracycline hydrochloride, can be used as a secondary standard
Certificates

Specifications

  • CAS No.:
    23313-80-6
  • Formula:
    C₂₂H₂₄N₂O₈.HCl
  • Description:
    4-Epitetracycline hydrochloride

Specifications

Related Information