Order Entry
China
ContactUsLinkComponent
Stachyose hydrate 97+%
Stachyose hydrate 97+%
Certificates
undefined
Stachyose hydrate 97+%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤12 % (K.F.)
  • Solubility:
    (5 % in water) Clear to slightly hazy colorless
  • HPLC:
    ≥97.0 % (ELSD)
  • Specific optical rotation:
    +129° to +140° (20°C, 589 nm) (c=4, H2O)

Specification Test Results

Related Information