Order Entry
China
ContactUsLinkComponent
Pipette set, BBP078 and BBP065
Pipette set, BBP078 and BBP065
Catalog # HACHLZP320
Supplier:  HACH
undefined
Pipette set, BBP078 and BBP065
Catalog # HACHLZP320
Supplier:  HACH

Specifications

  • Description:
    Incl. one BBP065 pipette (1,0 - 5,0 ml) with 75 tips and one BBP078 pipette (0,2 - 1,0 ml) with 100 tips
  • Seq:
    03
  • Pk:
    1 SET

Specifications

About this item

These variable volume single-channel, air-displacement pipettes feature outstanding accuracy with adjustments in 50 µl increments.

  • Robust design
  • Comfortable ergonomics
  • Slim-line tip ejection
  • Single-handed operation

Ideal for use with HACH TNTplus reagents.

Ordering information: Supplied with calibration report and maintenance tool.