Order Entry
China
ContactUsLinkComponent
Dry box gloves, neoprene, SP Bel-Art
Dry box gloves, neoprene, SP Bel-Art
Catalog # BELAH500290000
undefined
Dry box gloves, neoprene, SP Bel-Art
Catalog # BELAH500290000
Specifications
  • Ø:
    152 mm
  • Glove size:
    10
  • Colour:
    Black
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1 PAIR
Specifications
About this item
These neoprene gauntlets protect the user from penetration by liquid, gas or solid substances.
  • Seamless one-piece construction, made of bonded neoprene
  • Requires clamping rings to hold gloves in place