Order Entry
China
ContactUsLinkComponent
Sodium acetate in aqueous solution USP Test Solution (TS) 34
Sodium acetate in aqueous solution USP Test Solution (TS) 34
Catalog # VWRC85371.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium acetate in aqueous solution USP Test Solution (TS) 34
Catalog # VWRC85371.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :82.03 g/mol
Merck Index:12,08711
Melting Pt:324 °C
Storage Temperature:Ambient
EINECS:0
REACH:01-2119485123-42
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459