Skip to Header
Skip to Main Content
Skip to Footer
高级搜索
高级搜索
SDS
证书
制造商数据
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copper (II) acetate
Copper(II) acetate solution USP Test Solution (TS)
Copper(II) acetate solution USP Test Solution (TS)
Copper(II) acetate solution USP Test Solution (TS)
VWRC85341.180
:
VWR Chemicals
:
Copper(II) acetate solution USP Test Solution (TS)
VWRC85341.180
:
VWR Chemicals
:
Pk:
100 mL
Seq:
2440
:
:
181.64 g/mol
:
12,02690
:
115 °C
:
Ambient
:
0
:
9,III
:
142-71-2
:
CuCH₃CO₂
:
MFCD00008690