VWR® Transfer Pipettes, Graduated
Supplier: VWR BRAND PRIVATE LABEL USA
Certificates
About this item
These one-piece PE transfer pipettes feature reference (graduation) lines at 1 and 2 ml. Pipettes are designed with a small tip to ensure consistent reproduction of the drop size.
Ordering information: Non sterile pipettes are bulk packaged in two easy-to-use dispenser boxes of 500 each. Sterile pipettes are individually wrapped.