

Formic acid ≥90%, AnalaR NORMAPUR® for analysis of viscosity following DIN EN ISO 307
Catalog # VWRC85494.320
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (at filling):90.00 - 90.15 %
- Density (20/4) (at filling):1.2044 - 1.2047
- Acetic acid:≤ 0.2 %
- Conforms to ISO 307:Passes test
Specification Test Results
About this item
:
:46.03 g/mol
Merck Index:13,04265
Density:1.208 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:0
UN:1779
ADR:8 (3),II
:GHS02
:GHS05
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297