Order Entry
China
ContactUsLinkComponent
Pierce™ Disposable Columns
Pierce™ Disposable Columns
Catalog # PIER29920
undefined
Pierce™ Disposable Columns
Catalog # PIER29920
Specifications
  • Description:
    Disposable PS columns, for packing 0,5 - 2,0 ml bed volumes
  • Seq:
    01
  • Pk:
    100
Specifications
About this item
Empty polystyrene columns with matching frits and caps for use in a variety of resin based affinity or desalting techniques.
  • For benchtop gravity flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Compatible with most types of aqueous buffer eluents
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates