Order Entry
China
ContactUsLinkComponent
Automatic burettes, self-zeroing, SP Bel-Art
Catalog # BELAH379180112
undefined
Automatic burettes, self-zeroing, SP Bel-Art
Catalog # BELAH379180112
Specifications
  • Capacity:
    50 ml
  • Seq:
    02
  • Division:
    0,1 ml
  • Pk:
    1
Specifications
About this item
Self-zeroing glass burettes, Class B. Bottle, screw cap and round base made of PE.
  • Delivery end is fitted into a plastic push button stopcock which permits drop by drop delivery
  • Small screw clamp adjusts drop speed as needed for titration
  • PE filling tube extends from the top of the burette into the bottle reservoir
Burette have easy to read blue graduations against a white background. Complete with bottle and base.