Order Entry
China
ContactUsLinkComponent
VWR®, Powder packs for determination of nitrate, reagents for photometers
VWR®, Powder packs for determination of nitrate, reagents for photometers
Catalog # VWRC85612.620
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Powder packs for determination of nitrate, reagents for photometers
Catalog # VWRC85612.620
Supplier:  VWR Chemicals
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Pk:
    50 Tests
  • Description:
    Nitrate TUBES, RS
  • Range:
    0,2 - 30 mg/l NO₃- N
Specifications
About this item
VWR® Powder Pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners
RS = Powder pack reagent set