Order Entry
China
Orders LinkContactUsLinkComponent
Disposable samplers, DispoPicker
Disposable samplers, DispoPicker
Supplier:   BURKLE
Disposable samplers, DispoPicker
Supplier:  BURKLE
Certificates
Disposable samplers, DispoPicker
Disposable samplers, DispoPicker
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This disposable DispoPicker is used in the sampling of powders and small-grain bulk materials. Because the tip is closed, the sample material cannot trickle back out of the sample lance.
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
Ideally suited for cross-section samples.
Product Family Options
Product Information
  • Material
    Capacity
    Ø
    Length
    Sterile
    Availability
    Price
  • BURK5393-5540
    Disposable samplers
    Disposable samplers
    LaboPlast® HDPE
    60 ml
    21 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 20 件
    1 * 20 件
  • Disposable samplers
    SteriPlast® HDPE
    60 ml
    21 mm
    300 mm
    +
    Log in for availability
    Log in for availability
    1 * 20 件
    1 * 20 件
  • BURK5393-5550
    Disposable samplers
    Disposable samplers
    LaboPlast® HDPE
    100 ml
    21 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 20 件
    1 * 20 件
  • BURK5393-5551
    Disposable samplers
    Disposable samplers
    SteriPlast® HDPE
    100 ml
    21 mm
    500 mm
    +
    Log in for availability
    Log in for availability
    1 * 20 件
    1 * 20 件