pH Buffer solutions in bottles
pH Buffer solution, 7,01, ±0,01, Colourless, 25 °C, 500 mL
Supplier: HANNA
Jump to:
Product Details & Documents
Plastic bottle with a tamper-proof seal.
- Color coded label
Traceable to NIST standard reference materials.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:500 mL
- Reference temperature:25 °C
- Colour:Colourless
- Description:pH Buffer solution
- pH accuracy:±0,01
- pH value:7,01



