Order Entry
China
Orders LinkContactUsLinkComponent
Buffer solutions in sachets

Buffer solutions in sachets

Supplier:   HANNA
Buffer solutions in sachets
Supplier:  HANNA
Certificates
Buffer solutions in sachets

3 Options Available Below

Product Details & Documents

For calibration of pH meters.

  • Colour coded label for easy pH buffer value identification
  • Ideal for field use due to portability
  • Light block packaging prevents oxidation from UV light

Traceable to NIST standard reference materials.

Documents

Product Family Options

Product Information