Order Entry
China
ContactUsLinkComponent
Test kit, chlorine and pH, model CN-67
Test kit, chlorine and pH, model CN-67
Catalog # HACH1411100
Supplier:  HACH
CAS Number:  
undefined
Test kit, chlorine and pH, model CN-67
Catalog # HACH1411100
Supplier:  HACH
CAS Number:  

Specifications

  • Chemical method:
    DPD
  • Pk:
    1 KIT
  • Description:
    Two-parameter test kit, CN-67
  • Range:
    Chlorine: 0 - 3,5 mg/l
    pH: 6,5 - 8,5

Specifications

About this item

Maintaining safe water conditions and to protect against system corrosion by control of chlorine and pH. The free and total chlorine and pH test kit provides more accuracy than test kits using colour cubes or step-wise comparators.

Delivery: Includes colour discs and a comparator for monitoring chlorine and pH and required reagents and apparatus in a rugged carrying case for analysis in the field.