Order Entry
China
Orders LinkContactUsLinkComponent
Molybdenum standard solution

Molybdenum standard solution

Molybdenum standard solution (NIST), 10 mg/l as Mo, 100 mL
Catalog # HACH1418742
Supplier:   HACH
Molybdenum standard solution
Catalog # HACH1418742
Supplier:  HACH
CAS Number:  
Molybdenum standard solution
Molybdenum standard solution

Molybdenum standard solution (NIST), 10 mg/l as Mo, 100 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 毫升
Product Details & Documents

Standard solution for accuracy checks in molybdenum determinations.

Specifications

Specifications

  • Concentration:
    10 mg/l as Mo
  • Pk:
    100 mL
  • Description:
    Molybdenum standard solution (NIST)

Specifications