Order Entry
China
Orders LinkContactUsLinkComponent
Copper standard solution

Copper standard solution

Copper standard solution (NIST), 100 mg/l as Cu, 100 mL
Catalog # HACH12842
Supplier:   HACH
Copper standard solution
Catalog # HACH12842
Supplier:  HACH
CAS Number:  
Copper standard solution

Copper standard solution (NIST), 100 mg/l as Cu, 100 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 毫升
Product Details & Documents

Standard solution for accuracy checks in copper determinations.

Specifications

Specifications

  • Concentration:
    100 mg/l as Cu
  • Pk:
    100 mL
  • Description:
    Copper standard solution (NIST)

Specifications