Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
¥0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
China
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Accessories for visual comparator kit
Accessories for visual comparator kit
Accessories for visual comparator kit
Catalog # HACH990200
Supplier:
HACH
CAS Number:
Expand image
Accessories for visual comparator kit
Catalog # HACH990200
Supplier:
HACH
CAS Number:
Specifications
Pk:
1
Description:
DPD chlorine color disc
Specifications
About this item
Product Details
Product Details