

4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:108-10-1
:
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS02
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:= 98.5 %
- For Laboratory,Research,or Manufacturing Use:= 15
- Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water):≥98.5 %
- Color (APHA):≤15
- Residue after Evaporation:≤0.0050 %
- Titrable Acid (meq/g):≤0.002
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %