
Formamide ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # BAKRM522-06
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:230 mL
- Seq:2565
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:= 99.5 %
- For Laboratory,Research,or Manufacturing Use:2.0 - 3.0 °C
- Assay (HCONH₂) (by GC):≥99.5 %
- Freezing Point:2.0 - 3.0 °C
- Color (APHA):≤10
Specification Test Results
About this item
:
Merck Index:13,04261
Boiling Pt:210 °C (1013 hPa)
Melting Pt:2 °C
Density:1.1334 g/cm³ (20 °C)
Flash Pt:79 °C
:45.04 g/mol
Storage Temperature:Refrigerator
EINECS:200-842-0
:GHS08
CAS Number:75-12-7
Formula:HCONH₂
MDL Number:MFCD00007941