1,4-Butanediol, J.T.Baker®
Glass bottle, amber, narrow mouth, 500 mL
Supplier: Avantor
Jump to:
Product Details & Documents
Colorless, viscous liquid. Completely soluble in water.
Colorless, viscous liquid. Completely soluble in water.
Synonyms: 1,4-Dihydroxy butane, BDO, Tetramethylene glycol, 1,4-Butylene glycol
- CAS Number:110-63-4
- Formula:C₄H₁₀O₂
- MDL Number:MFCD00002968
- Molecular Weight:90.12 g/mol
- Boiling Pt:230 °C (1013 hPa)
- Melting Pt:19.5…20 °C
- Density:1.017 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-786-5
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Identification (by IR):Passes Test
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL



