


1-丙醇 ≥99.5% (通过GC), ChromAR® 用于液相色谱, 用于紫外分光光度法, Macron Fine Chemicals™
Catalog # MACR5351-08
Supplier: MACRON AVANTOR BRAND
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- 包装类型:Glass bottle, amber, narrow mouth
- 包装规格:4 L
- 顺序:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.5%
- Assay (CH₃(CH₂)₂OH) (by GC, corrected for water):≥99.5 %
- Residue (mg/L):≤3
- Water (H₂O)(by Karl Fischer titrn):≤0.08 %
- Optical Absorbance (1-cm path vs water) - 210 nm:≤1.00
- Optical Absorbance (1-cm path vs water) - 225 nm:≤0.50
- Optical Absorbance (1-cm path vs water) - 250 nm:≤0.05
- Optical Absorbance (1-cm path vs water) - 300 nm:≤0.005
- Optical Absorbance (1-cm path vs water) - 400 nm:≤0.005
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0,8053 g/cm³ (20 °C)
Flash Pt:15 °C
EINECS:200-746-9
UN:1274
ADR:3,III
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:71-23-8
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941