Order Entry
China
ContactUsLinkComponent
 
Toxic
Health
Chloroform ≥99.9% stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRH407-10
CAS Number:  
undefined
Toxic
Health
Chloroform ≥99.9% stabilised, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRH407-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.9 %
  • ACS - Assay (CHCl₃ including stabilizer) (by GC):
    ≥99.9 %
  • ACS - Residue after Evaporation:
    ≤0.0003 %
  • ACS - Water (H₂O)(by KF, coulometric):
    ≤0.03 %
  • Optical Absorbance (1-cm path vs water) - 245 nm:
    ≤1.00
  • Optical Absorbance (1-cm path vs water) - 255 nm:
    ≤0.15
  • Optical Absorbance (1-cm path vs water) - 260 nm:
    ≤0.05
  • Optical Absorbance (1-cm path vs water) - 270 nm:
    ≤0.02
  • Optical Absorbance (1-cm path vs water) - 290 - 400 nm:
    ≤0.01
  • Acetone, Aldehydes (as (CH₃)₂CO):
    Passes Test
  • Acid and Chloride:
    Passes Test
  • Color (APHA):
    ≤10
  • Free Chlorine:
    Passes Test
  • Lead (Pb):
    ≤0.05 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Optical Absorbance (1-cm path vs water) - Suitability for Use in Dithizone Tests:
    Passes Test

Specification Test Results

About this item

 :
 :119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
 :Stabilised with methanol 1 %
REACH:05-2114614906-45
 :GHS06
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826