Sodium acetate trihydrate 99.0-101.0% FCC, USP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:6131-90-4
:
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557
:136.08 g/mol
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Food GMP Manufactured Product:99.0 - 101.0 %
- Meets F.C.C. Requirements:Passes Test
- GMP Manufactured Product:= 0.035 %
- Meets U.S.P Requirements:= 10 ppm
- Preserve in tight containers, store at room temperature.:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Pharmaceutical Chemical:= 0.05 %
- Bulk Food Chemical:38.0 - 41.0 %
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤0.035 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Insoluble Matter:≤0.05 %
- USP - Loss on Drying:38.0 - 41.0 %
- USP - pH:7.5 - 9.2
- USP - Potassium Compounds:Passes Test
- USP - Sulfate (SO₄):≤0.005 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Alkalinity:≤0.05 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying:38.0 - 41.0 %
- FCC - Potassium Compounds:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR3462-01 spec_BAKR3462-07 spec_BAKR3462-09 spec_BAKR3462-R