

Butyl acetate ≥99.5% (by GC), PHOTREX® for spectrophotometry, J.T.Baker®
BAKR9191-03
: Avantor
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
- For Laboratory,Research,or Manufacturing Use:-
- ACS - Assay (CH₃COOCH₂CH₂CH₂CH₃) (by GC, corrected for water):≥ 99.5 %
- ACS - n-Butyl Alcohol (by GC):≤ 0.2 %
- ACS - Color (APHA):≤ 10
- ACS - Residue after Evaporation:≤ 0.0005 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Titrable Acid (meq/g):≤ 0.0016
- ACS - Water (by KF, volumetric):≤ 0.05 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400-320:≤ 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 300 nm:≤ 0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 275 nm:≤ 0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 263 nm:≤ 0.08
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 259 nm:≤ 0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 256 nm:≤ 0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 254 nm:≤ 1.0
:
:116.16 g/mol
:13,01534
:126 °C (1013 hPa)
:–76 °C
:0.875 g/cm³ (20 °C)
:25 °C
:Ambient
:204-658-1
:1123
:3,III
:GHS02
:GHS07
:123-86-4
:CH₃COO(CH₂)₃CH₃
:MFCD00009445