Order Entry
China
ContactUsLinkComponent
Adapters, reduction, SP Wilmad-LabGlass
Adapters, reduction, SP Wilmad-LabGlass
  LABGLG-1190-182
Adapters, reduction, SP Wilmad-LabGlass
  LABGLG-1190-182
  • Cone size:
    34/45 NS
  • Socket size:
    24/40 NS
  • Pk:
    1
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reducing adapter is used to allow connection of dissimilar NS joints. Outer joint at the top, inner joint at the bottom.