VWR®, Micro Spatulas
Supplier: VWR BRAND PRIVATE LABEL USA
Jump to:
About this item
Stainless steel. These micro spatulas are ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
- Rustproof material
- With non stick surfaces
- Mirror finish
They feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.
2 Options Available Below
Product Details & Documents
Stainless steel. These micro spatulas are ideal for small quantity applications such as sample transfer into test tubes or microcentrifuge tubes.
- Rustproof material
- With non stick surfaces
- Mirror finish
They feature one rounded end and one tapered end, allowing for a variety of transfer, mixing, or handling applications.