VWR®, Polyester Films for ELISA and General Incubation
Catalog # VWRU60941-062
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- Sterile:-
- Description:Polyester plate film with marking area
- Seq:03
- Thickness:50 µm
- Pk:100
Specifications
About this item
These 50 µm thick polyester films provide secure sealing around every well, not just a cover, thus minimising evaporation, preventing spillage and well-to-well contamination, and eliminating 'edge effects' in sensitive ELISA assays. Polyester films can also be used with tissue culture plates for short-term storage, incubation, and containment of biohazards.
- Functional temperature range from −40 to +120 °C
- Available sterile and non sterile
- Non pierceable
Strip films are designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard plates. Strip films are ideally suited for applications requiring rows to be selectively protected or accessed. Each strip film includes two removeable positioning tabs.
Each film L×D: 146×79 mm
Each strip film L×D: 103×19 mm