VWR®, Plate roller for securing films and foils to microplates
Catalog # VWRU60941-118
Supplier: VWR BRAND PRIVATE LABEL USA
Specifications
- Description:Plate roller
- Pk:1
Specifications
About this item
Soft rubber roller recommended for assuring tight and consistent application of adhesive films to microplates.